Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,6-Hexanetricarbonitrile 98.0+%, TCI America™
SDP

Catalog No. H150425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H150425G 25 g
H15045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H150425G Supplier TCI America Supplier No. H150425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1772-25-4
Molecular Formula C9H11N3
Molecular Weight (g/mol) 161.208
MDL Number MFCD00129792
InChI Key LNLFLMCWDHZINJ-UHFFFAOYSA-N
PubChem CID 15678
IUPAC Name hexane-1,3,6-tricarbonitrile
SMILES C(CC#N)CC(CCC#N)C#N

Specifications

Color Yellow
Boiling Point 200°C
Quantity 25 g
UN Number 3276
Formula Weight 161.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3,6-Hexanetricarbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.