Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Adamantanediol 98.0+%, TCI America™
SDP

Catalog No. A153125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A153125G 25 g
A15315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A153125G Supplier TCI America Supplier No. A153125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5001-18-3
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.236
MDL Number MFCD00154013
InChI Key MOLCWHCSXCKHAP-UHFFFAOYSA-N
Synonym 1,3-Dihydroxyadamantane
PubChem CID 573829
IUPAC Name adamantane-1,3-diol
SMILES C1C2CC3(CC1CC(C2)(C3)O)O

Specifications

Melting Point 258°C
Color White
Quantity 25 g
Formula Weight 168.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Adamantanediol
EINECSNumber (4)-1879
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.