Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Aminopropyl)imidazole 97.0+%, TCI America™
SDP

Catalog No. A1185100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1185100G 100 g
A118525G 25 g
A1185500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A1185100G Supplier TCI America Supplier No. A1185100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5036-48-6
Molecular Formula C6H11N3
Molecular Weight (g/mol) 125.18
MDL Number MFCD00009819
InChI Key KDHWOCLBMVSZPG-UHFFFAOYSA-N
Synonym 1-3-aminopropyl imidazole, n-3-aminopropyl imidazole, 1h-imidazole-1-propanamine, n-3-aminopropyl-imidazole, 3-1h-imidazol-1-yl propan-1-amine, 3-imidazol-1-ylpropylamine, 3-1h-imidazol-1-yl-1-propanamine, 1-3-aminopropyl-imidazole, 3-imidazol-1-yl propylamine, 3-imidazolylpropylamine
PubChem CID 78736
IUPAC Name 3-(1H-imidazol-1-yl)propan-1-amine
SMILES NCCCN1C=CN=C1

Specifications

Color Yellow
Boiling Point 117°C
Quantity 100 g
UN Number 2735
Formula Weight 125.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(3-Aminopropyl)imidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.