Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Benzodioxole, 99%

Catalog No. AAA141920B
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA141920B 1000 g
AAA1419218 50 g
AAA1419230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA141920B Supplier Thermo Scientific Chemicals Supplier No. A141920B
Add to Cart

CAS: 274-09-9 | C7H6O2 | 122.123 g/mol

1,3-Benzodioxole is useful in gemological stimulant detection. It is also used as a precursor for perfumes, photo initiators, agrochemicals and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Benzodioxole is useful in gemological stimulant detection. It is also used as a precursor for perfumes, photo initiators, agrochemicals and pharmaceuticals.

Solubility
Soluble in water (2g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 274-09-9
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.123
MDL Number MFCD00005818
InChI Key FTNJQNQLEGKTGD-UHFFFAOYSA-N
Synonym 1,2-methylenedioxybenzene, benzodioxole, benzo d 1,3 dioxole, 1,3-dioxaindan, 3,4-methylenedioxybenzene, 2h-1,3-benzodioxole, 1,2-methylenedioxy benzene, 1,3-dioxindan, methylenedioxybenzene, o-methylenedioxy benzene
PubChem CID 9229
ChEBI CHEBI:38732
IUPAC Name 1,3-benzodioxole
SMILES C1OC2=CC=CC=C2O1

Specifications

Melting Point -18°C
Density 1.185
Boiling Point 172°C to 173°C
Flash Point 55°C (131°F)
Odor Aromatic
Refractive Index 1.539
Quantity 1000 g
Beilstein 115506
Solubility Information Soluble in water (2g/L).
Formula Weight 122.12
Percent Purity 99%
Chemical Name or Material 1,3-Benzodioxole
Show More Show Less
Hazard Category H226-H302+H332
Hazard Statement GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 205-992-0
RTECSNumber DA5600000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.