Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Benzothiazol-5-amine, 97%, Thermo Scientific™

Catalog No. MO07036DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO07036DA 1 g Amber Glass Bottle
MO07036CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MO07036DA Supplier Thermo Scientific Chemicals Supplier No. MO07036DA

Chemical Identifiers

CAS 1123-93-9
Molecular Formula C7H6N2S
Molecular Weight (g/mol) 150.20
MDL Number MFCD04115282
InChI Key UJZYHMZRXGNDFB-UHFFFAOYSA-N
Synonym 5-benzothiazolamine, benzothiazol-5-ylamine, benzo d thiazol-5-amine, benzothiazol-5-amine, 5-amino-1,3-benzothiazole, 5-aminobenzothiazole, 1,3-benzothiazole-5-amine, 1,3-benzothiazol-5-ylamine, 5-amino-1,3-benzothiazol, 5-amino-benzothiazole
PubChem CID 70749
IUPAC Name 1,3-benzothiazol-5-amine
SMILES NC1=CC=C2SC=NC2=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 150.2
Percent Purity 95%
Chemical Name or Material 1, 3-Benzothiazol-5-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.