Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis[2-(4-hydroxyphenyl)-2-propyl]benzene 98.0+%, TCI America™
SDP

Catalog No. B2664250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2664250G 250 g
B266425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2664250G Supplier TCI America Supplier No. B2664250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13595-25-0
Molecular Formula C24H26O2
Molecular Weight (g/mol) 346.47
InChI Key PVFQHGDIOXNKIC-UHFFFAOYSA-N
Synonym Bisphenol M, 4,4′C-(1,3-Phenylenediisopropylidene)bisphenol
PubChem CID 3292100
IUPAC Name 4-[2-[3-[2-(4-hydroxyphenyl)propan-2-yl]phenyl]propan-2-yl]phenol
SMILES CC(C)(C1=CC=C(C=C1)O)C2=CC(=CC=C2)C(C)(C)C3=CC=C(C=C3)O

Specifications

Melting Point 138°C
Color White
Quantity 250 g
Formula Weight 346.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis[2-(4-hydroxyphenyl)-2-propyl]benzene
EINECSNumber (4)-1582
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.