Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis(2-isocyanato-2-propyl)benzene 97.0+%, TCI America™
SDP

Catalog No. B144225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B144225G 25 g
B1442500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B144225G Supplier TCI America Supplier No. B144225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2778-42-9
Molecular Formula C14H16N2O2
Molecular Weight (g/mol) 244.294
MDL Number MFCD00142493
InChI Key AZYRZNIYJDKRHO-UHFFFAOYSA-N
Synonym 1,3-Bis(1-isocyanato-1-methylethyl)benzene
PubChem CID 17719
IUPAC Name 1,3-bis(2-isocyanatopropan-2-yl)benzene
SMILES CC(C)(C1=CC(=CC=C1)C(C)(C)N=C=O)N=C=O

Specifications

Melting Point -10°C
Color Yellow
Boiling Point 106°C
Quantity 25 g
UN Number 2810
Formula Weight 244.29
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Bis(2-isocyanato-2-propyl)benzene
EINECSNumber (3)-3908
RTECSNumber CY8480000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.