Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis[4-(trifluoromethyl)phenyl]urea 98.0+%, TCI America™
SDP

Catalog No. B45181G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45181G 1 g
B4518200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45181G Supplier TCI America Supplier No. B45181G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1960-88-9
Molecular Formula C15H10F6N2O
Molecular Weight (g/mol) 348.248
MDL Number MFCD01908917
InChI Key PEXQIAHZLFTVJU-UHFFFAOYSA-N
Synonym 1,3-bis 4-trifluoromethyl phenyl urea, maybridge4_001077, 1,3-bis 4-trifluoromethylphenyl urea, n,n'-bis-4-trifluoromethylphenyl-urea, urea, n,n'-bis 4-trifluoromethyl phenyl
PubChem CID 472928
IUPAC Name 1,3-bis[4-(trifluoromethyl)phenyl]urea
SMILES C1=CC(=CC=C1C(F)(F)F)NC(=O)NC2=CC=C(C=C2)C(F)(F)F

Specifications

Melting Point 227°C
Color White
Quantity 1 g
Formula Weight 348.25
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis[4-(trifluoromethyl)phenyl]urea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.