Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[1,3-Bis(diphenylphosphino)propane]nickel(II) Dichloride 98.0+%, TCI America™
SDP

Catalog No. B131325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B131325G 25 g
B13135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B131325G Supplier TCI America Supplier No. B131325G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 15629-92-2
Molecular Formula C27H26Cl2NiP2
Molecular Weight (g/mol) 542.04
MDL Number MFCD00015318
InChI Key REQVOKPBMSCTNI-UHFFFAOYSA-L
Synonym [1,3-Bis(diphenylphosphino)propane]dichloronickel(II), Dichloro[1,3-bis(diphenylphosphino)propane]nickel(II)
PubChem CID 131675641
IUPAC Name [3-(diphenylphosphanyl)propyl]diphenylphosphane; dichloronickelbis(ylium)
SMILES Cl[Ni++]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 213°C
Quantity 25 g
Formula Weight 542.05
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material [1,3-Bis(diphenylphosphino)propane]nickel(II) Dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.