Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Butadiene in Nitrogen, 10 ppm, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111003976 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
14 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-3976 14 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-3976 Supplier MilliporeSigma Supelco Supplier No. 303405

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Specifications

Packaging Cylinder
Physical Form Gas
Concentration 10 ppm
Quantity 14 L
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.