Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-1,3-Butanediol, 99%, extra pure

Catalog No. AC107620010
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107620010 1 L Glass bottle
AC107622500 250 mL Glass bottle
AC107620025 2.5 L Glass bottle
AC107620100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC107620010 Supplier Thermo Scientific Chemicals Supplier No. 107620010
Add to Cart

CAS: 107-88-0 | C4H10O2 | 90.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-88-0
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00004554
InChI Key PUPZLCDOIYMWBV-UHFFFAOYSA-N
Synonym 1,3-butanediol, 1,3-butylene glycol, 1,3-dihydroxybutane, methyltrimethylene glycol, 1,3-butandiol, 1-methyl-1,3-propanediol, beta-butylene glycol, 1,3 butylene glycol, 1,3-butylenglykol, rs-1,3-butandiol
PubChem CID 7896
ChEBI CHEBI:52683
IUPAC Name butane-1,3-diol
SMILES CC(CCO)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -54.0°C
Color Brown to Colorless
Density 1.0100g/mL
Boiling Point 203.0°C to 204.0°C
Flash Point 109°C
Ignition Residue 0.05% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4385 to 1.4405
Linear Formula CH3CH(OH)CH2CH2OH
Quantity 1 L
Beilstein 01, 477
Specific Gravity 1.01
Merck Index 15, 1570
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone, methyl ethyl ketone, ethanol, slightly soluble in ether, practically insoluble in aliphatic hydrocarbons,, benzene, toluene and ccl4.
Viscosity 24.6 mm2/s (50°C)
Formula Weight 90.12
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material (±)-1,3-Butanediol
Show More Show Less
EINECSNumber 203-529-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.