Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Chlorobenzyl)piperazine, 98%, Thermo Scientific™

Catalog No. AAH5540906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5540906 5 g
AAH5540903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5540906 Supplier Thermo Scientific Chemicals Supplier No. H5540906

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23145-91-7
Molecular Formula C11H15ClN2
Molecular Weight (g/mol) 210.705
MDL Number MFCD02048053
InChI Key JTEQMTYOCBFLNH-UHFFFAOYSA-N
Synonym 1-3-chlorobenzyl piperazine, 1-3-chlorophenyl methyl piperazine, 1-3-chloro-benzyl-piperazine, 3-chlorophenyl methyl piperazine, 1-3-chlorophenyl methyl piperazine hydrochloride, acmc-1cngg, 1-3-chlorobenzyl-piperazine, 4-3-chlorophenyl methylpiperazine, piperazine,1-3-chlorophenyl methyl
PubChem CID 764727
IUPAC Name 1-[(3-chlorophenyl)methyl]piperazine
SMILES C1CN(CCN1)CC2=CC(=CC=C2)Cl

Specifications

Color Colorless to Yellow
Boiling Point 120°C to 123°C (1.5 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN2735
Formula Weight 210.71
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-(3-Chlorobenzyl)piperazine
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.