Learn More
Sigma Organic Chemistry 1-3-Chloropropyl -1 3-d | 250mg | 62780-89-6 | MFCD00044896
Supplier: Sigma Organic Chemistry CDS008063250MG

1-3-Chloropropyl -1 3-d , 250mg
About this Item:
CAS #: 62780-89-6
MDL #: MFCD00044896
Molecular Weight: 210.66
UNSPSC Code: 12352200
Chemical Formula: C10H11ClN2O
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.