Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Chloropropyl)piperidine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. C29221G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29221G 1 g
C29225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29221G Supplier TCI America Supplier No. C29221G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5472-49-1
Molecular Formula C8H17Cl2N
Molecular Weight (g/mol) 198.131
MDL Number MFCD00012838
InChI Key OBOBUDMMFXRNDO-UHFFFAOYSA-N
Synonym 1-3-chloropropyl piperidine hydrochloride, n-3-chloropropyl piperidine hydrochloride, piperidine, 1-3-chloropropyl-, hydrochloride, piperidinopropyl chloride, hydrochloride, 3-piperidinopropyl chloride hydrochloride, 1-3-chloropropyl piperidine monohydrochloride, 1-3-chloropropyl piperidine hcl, n-3-chloropropyl-piperidine hydrochloride, 1-3-chloropropyl piperidine hydrochloride 1:1, acmc-1ak9t
PubChem CID 79624
IUPAC Name 1-(3-chloropropyl)piperidine;hydrochloride
SMILES C1CCN(CC1)CCCCl.Cl

Specifications

Melting Point 218°C
Color White
Quantity 1 g
Formula Weight 198.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(3-Chloropropyl)piperidine Hydrochloride
RTECSNumber TM6498600
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.