Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Di(2-pyridyl)propane 97.0+%, TCI America™
SDP

Catalog No. D541525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D541525G 25 g
D5415500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D541525G Supplier TCI America Supplier No. D541525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15937-81-2
Molecular Formula C13H14N2
Molecular Weight (g/mol) 198.269
InChI Key UHPWYBATPHCMGD-UHFFFAOYSA-N
PubChem CID 322131
IUPAC Name 2-(3-pyridin-2-ylpropyl)pyridine
SMILES C1=CC=NC(=C1)CCCC2=CC=CC=N2

Specifications

Boiling Point 117°C
Quantity 25 g
Formula Weight 198.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Di(2-pyridyl)propane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.