Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Di-9-carbazolylbenzene (purified by sublimation) 98.0+%, TCI America™
SDP

Catalog No. D40871G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D40871G Supplier TCI America Supplier No. D40871G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 550378-78-4
Molecular Formula C30H20N2
Molecular Weight (g/mol) 408.504
MDL Number MFCD09836836
InChI Key MZYDBGLUVPLRKR-UHFFFAOYSA-N
Synonym 1,3-Bis(carbazol-9-yl)benzene
PubChem CID 22020377
IUPAC Name 9-(3-carbazol-9-ylphenyl)carbazole
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC(=CC=C4)N5C6=CC=CC=C6C7=CC=CC=C75

Specifications

Melting Point 184°C
Color White
Quantity 1 g
Formula Weight 408.50
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Di-9-carbazolylbenzene (purified by sublimation)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.