Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diacetoxybenzene, 97%

Catalog No. AAA115470B
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA115470B 1000 g
AAA1154718 50 g
AAA1154730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA115470B Supplier Thermo Scientific Chemicals Supplier No. A115470B
Add to Cart

CAS: 108-58-7 | C10H10O4 | 194.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-58-7
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00008701
InChI Key STOUHHBZBQBYHH-UHFFFAOYSA-N
Synonym 1,3-diacetoxybenzene, resorcinol diacetate, 1,3-benzenediol, diacetate, m-phenylenediacetate, resorcinol, diacetate, m-phenylene di acetate, 1,3-dihydroxybenzene diacetate, dihydroxybenzene diacetate, 1,3-phenylene diacetate, 1,3-benzenediol, 1,3-diacetate
PubChem CID 7942
IUPAC Name (3-acetyloxyphenyl) acetate
SMILES CC(=O)OC1=CC(=CC=C1)OC(=O)C

Specifications

Density 1.179
Boiling Point 278°C
Flash Point >110°C (230°F)
Refractive Index 1.503
Quantity 1000 g
Beilstein 1875007
Formula Weight 194.19
Percent Purity 98%
Chemical Name or Material 1,3-Diacetoxybenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 203-596-2
RTECSNumber VH0580000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.