Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diaminopropane, Spectrum™ Chemical
SDP

Catalog No. 18620602 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
200 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-602 200 L
18-620-601 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-620-602 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D2049200LTBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Synonym: 1,3-Propanediamine, Trimethylenediamine, 1,3-Diamine-Propane

Chemical Identifiers

CAS 109-76-2

Specifications

Boiling Point 140°C (284°F)
Packaging Amber Bottle
Quantity 200 L
Molecular Weight (g/mol) 74.13
Percent Purity 99%
Grade Ungraded
Physical Form Liquid
Chemical Name or Material 1,3-Diaminopropane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.