Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dibenzoyloxybenzene 98.0+%, TCI America™
SDP

Catalog No. D520725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D520725G 25 g
D52075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D520725G Supplier TCI America Supplier No. D520725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 94-01-9
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00016576
InChI Key SUQGLJRNDJRARS-UHFFFAOYSA-N
Synonym 1,3-dibenzoyloxybenzene, resorcinol dibenzoate, 1,3-benzenediol, dibenzoate, 1,3-phenylene dibenzoate, m-phenylene dibenzoate, resorcinol, dibenzoate, 1,3-bis benzoyloxy benzene, 3-benzoyloxy phenyl benzoate, 1,3-benzenediol, 1,3-dibenzoate, 1,3-phenylenedibenzoate
PubChem CID 66742
IUPAC Name 3-(benzoyloxy)phenyl benzoate
SMILES O=C(OC1=CC(OC(=O)C2=CC=CC=C2)=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 118°C
Color White
Quantity 25 g
Formula Weight 318.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dibenzoyloxybenzene
RTECSNumber VH0590000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.