Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dibromo-2-iodobenzene 98.0+%, TCI America™
SDP

Catalog No. D42941G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42941G 1 g
D42945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42941G Supplier TCI America Supplier No. D42941G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19821-80-8
Molecular Formula C6H3Br2I
Molecular Weight (g/mol) 361.80
MDL Number MFCD00192666
InChI Key OIFRMZVTJQAPIF-UHFFFAOYSA-N
PubChem CID 4403794
IUPAC Name 1,3-dibromo-2-iodobenzene
SMILES BrC1=CC=CC(Br)=C1I

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 1 g
Formula Weight 361.80
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dibromo-2-iodobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.