Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dibromo-4-nitrobenzene, 98%, Thermo Scientific™

Catalog No. AAH6173803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6173803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6173803 Supplier Thermo Scientific Chemicals Supplier No. H6173803

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51686-78-3
Molecular Formula C6H3Br2NO2
Molecular Weight (g/mol) 280.903
MDL Number MFCD02660416
InChI Key DXRVYZGVVFZCFP-UHFFFAOYSA-N
PubChem CID 12315646
IUPAC Name 2,4-dibromo-1-nitrobenzene
SMILES C1=CC(=C(C=C1Br)Br)[N+](=O)[O-]

Specifications

Color Yellow
Quantity 1 g
UN Number UN3459
Synonym 2,4-dibromonitrobenzene, benzene, 2,4-dibromo-1-nitro, 2,4-dibromo-1-nitro-benzene, 1,3-dibromo-4-nitrobenzene, acmc-209kve, 2,4-dibromo-nitrobenzene, ksc594m9r, benzene,2,4-dibromo-1-nitro, 2,4-bis bromanyl-1-nitro-benzene, 2,4-dibromo-1-nitrobenzene;,3-dibromo-4-nitrobenzene
Formula Weight 280.9
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dibromo-4-nitrobenzene
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITROBROMOBENZENES, SOLID
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.