Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dibromo-5-chlorobenzene 96.0+%, TCI America™
SDP

Catalog No. D346525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D346525G 25 g
D34655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D346525G Supplier TCI America Supplier No. D346525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14862-52-3
Molecular Formula C6H3Br2Cl
Molecular Weight (g/mol) 270.348
MDL Number MFCD00070765
InChI Key FNKCOUREFBNNHG-UHFFFAOYSA-N
Synonym 3,5-dibromochlorobenzene, 1-chloro-3,5-dibromobenzene, benzene, 1,3-dibromo-5-chloro, pubchem3634, ambsclk-224, intermediates-zcf02139, ksc181m3d, 1,3-dibromo-5-chloro-benzene, tpc-i027, fnkcourefbnnhg-uhfffaoysa
PubChem CID 84676
IUPAC Name 1,3-dibromo-5-chlorobenzene
SMILES C1=C(C=C(C=C1Br)Br)Cl

Specifications

Melting Point 96°C
Color White-Yellow
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Boiling Point 256°C
Formula Weight 270.35
Percent Purity ≥96.0% (GC)
Chemical Name or Material 1,3-Dibromo-5-chlorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.