Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dibromo-5-tetradecylbenzene 97.0+%, TCI America™
SDP

Catalog No. D52581G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52581G 1 g
D52585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52581G Supplier TCI America Supplier No. D52581G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 157761-91-6
Molecular Formula C20H32Br2
Molecular Weight (g/mol) 432.284
InChI Key DEEYQDRVVZEMDY-UHFFFAOYSA-N
PubChem CID 101945988
IUPAC Name 1,3-dibromo-5-tetradecylbenzene
SMILES CCCCCCCCCCCCCCC1=CC(=CC(=C1)Br)Br

Specifications

Melting Point 37°C
Boiling Point 210°C
Quantity 1 g
Formula Weight 432.28
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dibromo-5-tetradecylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.