Learn More
1,3-Dibromobenzene, 97+%
Description
1,3-Dibromobenzene is used in Suzuki coupling reaction for the preparation of [n]metacyclophanes. It is used to prepare 5-(3-bromo-phenyl)-2,4-di-tert-butoxy-pyrimidine using Pd(0) (PPh3)4 as a reagent. Further, it undergoes solid-supported potassium fluoride-aluminum oxide palladium-catalyzed polyarylation reaction with phenyl boronic acid to get conjugated polyaryls.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,3-Dibromobenzene is used in Suzuki coupling reaction for the preparation of [n]metacyclophanes. It is used to prepare 5-(3-bromo-phenyl)-2,4-di-tert-butoxy-pyrimidine using Pd(0) (PPh3)4 as a reagent. Further, it undergoes solid-supported potassium fluoride-aluminum oxide palladium-catalyzed polyarylation reaction with phenyl boronic acid to get conjugated polyaryls.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | -7°C |
| Density | 1.955 |
| Boiling Point | 219°C to 220°C |
| Flash Point | 93°C (199°F) |
| Odor | Odorless |
| Assay Percent Range | 97% |
| Refractive Index | 1.608 |
| Quantity | 100 g |
| UN Number | UN3082 |
| Beilstein | 1904538 |
| Show More |
Safety and Handling
| Hazard Category | H227-H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H227 Causes skin irritation. Causes serious eye irritation. Combustible liquid. |
| Precautionary Statement | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S. |
| EINECSNumber | 203-574-2 |
| RTECSNumber | CZ1790000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.