Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dichloro-2-propanol 98.0+%, TCI America™
SDP

Catalog No. D0402500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0402500G 500 g
D040225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0402500G Supplier TCI America Supplier No. D0402500G

Chemical Identifiers

CAS 96-23-1
Molecular Formula C3H6Cl2O
Molecular Weight (g/mol) 128.98
MDL Number MFCD00000951
InChI Key DEWLEGDTCGBNGU-UHFFFAOYSA-N
Synonym 1,3-dichloro-2-propanol, dichlorohydrin, 1,3-dichloroisopropanol, 1,3-dichlorohydrin, glycerol 1,3-dichlorohydrin, 2-propanol, 1,3-dichloro, enodrin, 1,3-dichloro-2-hydroxypropane, 1,3-dichloroisopropyl alcohol, sym-glycerol dichlorohydrin
PubChem CID 7289
ChEBI CHEBI:18917
IUPAC Name 1,3-dichloropropan-2-ol
SMILES OC(CCl)CCl

Specifications

Melting Point -4°C
Color Yellow
Boiling Point 174°C
Quantity 500 g
UN Number 2750
Formula Weight 128.98
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dichloro-2-propanol
EINECSNumber (2)-2002
RTECSNumber UB1400000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.