Learn More
1,3-Dichloro-5,5-dimethylhydantoin, 98%
Description
Dantoin(R) DCDMH is an industrial bleaching agent based on dimethyl hydantoin. This product finds application wherever solid bleach is desired. It is used as a mild chlorinating agent in organic synthesis.It is also used as a sanitizer in swimming pools, laundries restaurants, cutting oils and pharmaceutical industry as a general cleaner. Other uses include it is been used as a preventer and catalyst in polymer industry, used to emboss or texturize resinous sheet preparations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dantoin(R) DCDMH is an industrial bleaching agent based on dimethyl hydantoin. This product finds application wherever solid bleach is desired. It is used as a mild chlorinating agent in organic synthesis.It is also used as a sanitizer in swimming pools, laundries restaurants, cutting oils and pharmaceutical industry as a general cleaner. Other uses include it is been used as a preventer and catalyst in polymer industry, used to emboss or texturize resinous sheet preparations.
Solubility
Soluble in water 0.21 g/100 mL (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with reducing agents, acids, strong bases, organic materials, powdered metals. Moisture, light, air sensitive. Stable, but a strong oxidizer.
Specifications
Specifications
| Melting Point | 130°C to 134°C |
| Density | 1.5 |
| Flash Point | 174°C (345°F) |
| Quantity | 2500 g |
| UN Number | UN1479 |
| Beilstein | 146013 |
| Sensitivity | Moisture sensitive |
| Merck Index | 14,3065 |
| Solubility Information | Soluble in water 0.21g/100 mL (25°C). |
| Formula Weight | 197.02 |
| Show More |
Safety and Handling
| Hazard Category | H272-H302-H314-H317 |
| Hazard Statement | GHS H Statement H272-H301-H314-H318 May intensify fire; oxidizer. Toxic if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P210-P220-P221-P260-P264b-P270-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S. |
| EINECSNumber | 204-258-7 |
| RTECSNumber | MU0700000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.