Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dichloro-5-(chloromethyl)benzene, 97%, Thermo Scientific™

Catalog No. SPB00444EA
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SPB00444EA 10 g Amber Glass Bottle
SPB00444DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SPB00444EA Supplier Thermo Scientific Chemicals Supplier No. SPB00444EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3290-06-0
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.467
InChI Key ZFLRKAMKGYNFPH-UHFFFAOYSA-N
Synonym 3,5-dichlorobenzyl chloride, 1,3-dichloro-5-chloromethyl benzene, benzene, 1,3-dichloro-5-chloromethyl, 3,5-dichlorobenzylchloride, unii-c076i19eql, 5-chloromethyl-1,3-dichlorobenzene, pubchem16695, alpha,3,5-trichlorotoluene, 3,4-dichloro benzyl chloride, ksc495g8l
PubChem CID 137880
IUPAC Name 1,3-dichloro-5-(chloromethyl)benzene
SMILES C1=C(C=C(C=C1Cl)Cl)CCl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula Cl2C6H3CH2Cl
Quantity 10 g
Formula Weight 195.48
Percent Purity 97%
Chemical Name or Material 1, 3-Dichloro-5-(chloromethyl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.