Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dichlorobenzene Solution, 1 Analyte, 100μg/mL in Methanol, Ultra Scientific

Catalog No. USHC1201
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-HC-120-1 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-HC-120-1 Supplier Agilent Technologies Supplier No. HC1201
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 541-73-1 , 67-56-1
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 146.998
InChI Key ZPQOPVIELGIULI-UHFFFAOYSA-N
PubChem CID 10943
ChEBI CHEBI:36693
IUPAC Name 1,3-dichlorobenzene
SMILES C1=CC(=CC(=C1)Cl)Cl

Specifications

Physical Form Liquid
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material 1, 3-Dichlorobenzene, 100μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.