Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Acrylamide, 99+%, for biochemistry, electrophoresis grade

Catalog No. AC164850025
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164850025 2.5 kg Plastic Bottle
AC164851000 100 g Plastic Bottle
AC164855000 500 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC164850025 Supplier Thermo Scientific Chemicals Supplier No. 164850025
Only null left
Add to Cart

Acrylamide, CAS # 79-06-1, is an organic amide compound used as a monomer in the synthesis of polyacrylamides. | CAS: 79-06-1 | C3H5NO | 71.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Acrylamide is an organic amide compound mainly used to as a monomer in the synthesis of polyacrylamide
• Acrylamide is formed in foods containing carbohydrates in its composition, mainly when these foods are fried, grilled or baked

Applications

• In laboratory experiments acrylamide participates in chemical reactions as an intermediate in the preparation of N-methylol acrylamide and N-butoxyacrylamide compounds
• For molecular usage, this compound can be used as a carrier and is involved in the precipitation of small amounts of DNA
• In molecular experiments it is used in gel electrophoresis, including western blot, and SDS-PAGE, to separate proteins and macromolecules based on their size and charge

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-06-1
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
Synonym Ethylenecarboxamide, 2-Propenamide
PubChem CID 6579
ChEBI CHEBI:28619
IUPAC Name prop-2-enamide
SMILES NC(=O)C=C

Specifications

Physical Form Crystals or Crystalline Powder
Acrylic Acid 0.002% max.
Boiling Point 125.0°C (25.0 mmHg)
CAS Min % 99.0
Melting Point 82.0°C to 86.0°C
CAS Max % 100.0
Conductivity 5 umhos (40% max. aqueous solution)
Density 1.1220g/mL
Color White
pH 5.5 to 6.5 (10% soln., 0.1 M NaCl, 25°C)
Quantity 2.5 kg
Flash Point 138°C
Infrared Spectrum Authentic
Packaging Plastic Bottle
Assay Percent Range 99+%
Linear Formula H2C=CHCONH2
Beilstein 02, 400
Merck Index 15, 120
Solubility Information Solubility in water: 21.5g/L (30°C). Other solubilities: solubilities in g/100mL solvent (30°C):, methanol 155; ethanol 86.2; acetone 63.1;, ethyl acetate 12.6; chloroform 2.66;, benzene 0.346; heptane 0.0068, soluble in ether
Formula Weight 71.08
Percent Purity ≥99%
Grade Biochemical
Chemical Name or Material Acrylamide, 99+%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
Toxic if swallowed.
May cause an allergic skin reaction.
Harmful if inhaled.
May cause cancer.
May cause genetic defects.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-173-7
RTECSNumber AS3325000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.