Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Difluoropropane, 97%, Thermo Scientific™

Catalog No. AAL1235106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1235106 5 g
AAL1235114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1235106 Supplier Thermo Scientific Chemicals Supplier No. L1235106

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 462-39-5
Molecular Formula C3H6F2
Molecular Weight (g/mol) 80.08
MDL Number MFCD00040969
InChI Key OOLOYCGJRJFTPM-UHFFFAOYSA-N
Synonym propane, 1,3-difluoro, acmc-1aiey, 1,3-bis fluoranyl propane
PubChem CID 68041
IUPAC Name 1,3-difluoropropane
SMILES FCCCF

Specifications

Density 1.006
Boiling Point 40°C to 42°C
Refractive Index 1.321
Linear Formula FCH2CH2CH2F
Quantity 5 g
Beilstein 1696999
Formula Weight 80.08
Percent Purity 97%
Chemical Name or Material 1,3-Difluoropropane
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.