Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dihydroxynaphthalene, For Spectrophotometric Det. of Glucuronic Acid According to Tollens, ≥97.0%, MilliporeSigma™ Supelco™

Catalog No. 111012949 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2949 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2949 Supplier MilliporeSigma Supelco Supplier No. 706505G
Add to Cart
Add to Cart

Chemical Identifiers

MDL Number MFCD00003965
Synonym 1,3-Naphthalenediol; Naphthoresorcinol

Specifications

Melting Point 122°C to 125°C; 123°C to 125°C
CAS 132-86-5
Linear Formula C10H6(OH)2
Quantity 5 g
Formula Weight 160.17
Percent Purity ≥97.0% (HPLC); ≥97.0%
RTECSNumber QJ4725000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.