Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diiodo-5,5-dimethylhydantoin 97.0+%, TCI America™
SDP

Catalog No. D365725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D365725G 25 g
D36575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D365725G Supplier TCI America Supplier No. D365725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2232-12-4
Molecular Formula C5H6I2N2O2
Molecular Weight (g/mol) 379.92
MDL Number MFCD00020867
InChI Key RDZHCKRAHUPIFK-UHFFFAOYSA-N
Synonym DIH
PubChem CID 200534
IUPAC Name 1,3-diiodo-5,5-dimethylimidazolidine-2,4-dione
SMILES CC1(C(=O)N(C(=O)N1I)I)C

Specifications

Melting Point 198°C
Quantity 25 g
UN Number 3085
Formula Weight 379.92
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diiodo-5,5-dimethylhydantoin
EINECSNumber (5)-7009
RTECSNumber MU0970000
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.