Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diisopropylimidazolium chloride, 97+%

Catalog No. AAH2779803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2779803 1 g
AAH2779806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2779803 Supplier Thermo Scientific Chemicals Supplier No. H2779803
Add to Cart

CAS: 139143-09-2 | C9H17ClN2 | 188.70 g/mol

Ligand for ruthenium-catalyzed greener amide bond formation by dehydrogenative coupling of amines and alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ligand for ruthenium-catalyzed greener amide bond formation by dehydrogenative coupling of amines and alcohols.

Solubility
Soluble in water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139143-09-2
Molecular Formula C9H17ClN2
Molecular Weight (g/mol) 188.70
MDL Number MFCD03840577
InChI Key DOFXKPAOJLLPII-UHFFFAOYSA-M
Synonym 1,3-diisopropylimidazolium chloride, 1,3-di-i-propylimidazolium chloride, 1,3-diisopropyl-1h-imidazol-3-ium chloride, 1,3-diisopropylimidazol-1-ium chloride, diic, acmc-20ap45, n,n'-isopropyl imidazolium chloride, 1h-imidazolium,1,3-bis 1-methylethyl-, chloride 1:1, n,n inverted exclamation marka-isopropyl imidazolium chloride
PubChem CID 11229242
IUPAC Name 1,3-di(propan-2-yl)imidazol-1-ium;chloride
SMILES [Cl-].CC(C)N1C=C[N+](=C1)C(C)C

Specifications

Melting Point 182°C to 186°C
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
Formula Weight 188.69
Percent Purity ≥97%
Chemical Name or Material 1,3-Diisopropylimidazolium chloride
Hazard Category H302+H312+H332-H315-H319-H320-H335-H336
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P330+P331-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.