Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dimethyl-2-thiohydantoin, 95%, Thermo Scientific™

Catalog No. AAH6338403
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6338403 1 g
AAH63384MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6338403 Supplier Thermo Scientific Chemicals Supplier No. H6338403

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1801-62-3
Molecular Formula C5H8N2OS
Molecular Weight (g/mol) 144.192
MDL Number MFCD00186242
InChI Key GKWNLFPHQCFFQJ-UHFFFAOYSA-N
Synonym 1,3-dimethyl-2-thiohydantoin, 1,3-dimethyl-2-thioxoimidazolidin-4-one, n,n'-dimethylthiohydantoin, 1,3-dimethyl-2-thioxo-4-imidazolidinone, 4-imidazolidinone,1,3-dimethyl-2-thioxo, acmc-1c9l4, 1,3-dimethyl-2-thioxo-1,3-diazolidin-4-one
PubChem CID 3722203
IUPAC Name 1,3-dimethyl-2-sulfanylideneimidazolidin-4-one
SMILES CN1CC(=O)N(C1=S)C

Specifications

Color Orange
Quantity 1 g
Formula Weight 144.19
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 1,3-Dimethyl-2-thiohydantoin
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.