Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-[3-(Dimethylamino)propyl]-3-ethylurea, 97%

Catalog No. AAH6384506
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6384506 5 g
AAH63845MD 250 mg
AAH6384503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6384506 Supplier Thermo Scientific Chemicals Supplier No. H6384506
Add to Cart

CAS: 32897-26-0 | C8H19N3O | 173.26 g/mol

1-[3-(Dimethylamino)propyl]-3-ethylurea is used in the preparation of cabergoline, which is a dopamine receptor used to treat Parkinsons syndrom.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-[3-(Dimethylamino)propyl]-3-ethylurea is used in the preparation of cabergoline, which is a dopamine receptor used to treat Parkinsons syndrom.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32897-26-0
Molecular Formula C8H19N3O
Molecular Weight (g/mol) 173.26
MDL Number MFCD00027752
InChI Key NGJUYARYEXGDNN-UHFFFAOYSA-N
Synonym 1-3-dimethylamino propyl-3-ethylurea, 1-ethyl-3 3-dimethylamino urea, 1-ethyl-3-3-dimethylaminopropyl urea, urea, n-3-dimethylamino propyl-n'-ethyl, 3-3-dimethylamino propyl-1-ethylurea, 1-3-dimethylamino propyl-3-ethyl isourea, 1-3-dimethylaminopropyl-3-ethylurea, 3-ethyl amino hydroxy methylene amino-n,n-dimethylpropan-1-amine
PubChem CID 4196313
IUPAC Name 1-[3-(dimethylamino)propyl]-3-ethylurea
SMILES CCNC(=O)NCCCN(C)C

Specifications

Linear Formula CH3CH2NHCONH(CH2)3N(CH3)2
Quantity 5 g
Beilstein 4961600
Solubility Information Soluble in water.
Formula Weight 173.26
Percent Purity 97%
Chemical Name or Material 1-[3-(Dimethylamino)propyl]-3-ethylurea
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.