Learn More
1,3-Dimethylbarbituric acid, 99% (dry wt.), water <6%
Description
1,3-Dimethylbarbituric acid is used as a catalyst in the Knoevenagel condensation of a series of aromatic aldehydes. It is also used in the synthesis of 5-aryl-6-(alkyl- or aryl-amino)-1,3-dimethylfuro [2,3-d]pyrimidine derivatives and enantioselective synthesis of isochromene pyrimidinedione derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 121°C to 123°C |
| Quantity | 50 g |
| Physical Form | Crystalline Powder |
| Beilstein | 139810 |
| Formula Weight | 156.14 |
| Percent Purity | 99% |
| Concentration or Composition (by Analyte or Components) | water <6% |
| Assay | (dry weight) |
| Chemical Name or Material | 1,3-Dimethylbarbituric acid |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement H301-H318 Toxic if swallowed. Causes serious eye damage. |
| Precautionary Statement | GHS P Statement P280-P301+P310-P305+P351+P338-P310-P405-P501a Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| TSCA | TSCA |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.