Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dimethylimidazolium Chloride 98.0+%, TCI America™
SDP

Catalog No. D334125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D334125G 25 g
D33415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D334125G Supplier TCI America Supplier No. D334125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 79917-88-7
Molecular Formula C5H9ClN2
Molecular Weight (g/mol) 132.591
InChI Key IIJSFQFJZAEKHB-UHFFFAOYSA-M
PubChem CID 11062432
IUPAC Name 1,3-dimethylimidazol-1-ium;chloride
SMILES CN1C=C[N+](=C1)C.[Cl-]

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 25 g
Formula Weight 132.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dimethylimidazolium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.