Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dimethylimidazolium dimethyl phosphate, 98%

Catalog No. p-7030231
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2778806 5 g
AAH2778818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2778806 Supplier Thermo Scientific Chemicals Supplier No. H2778806
Only null left
Add to Cart

CAS: 654058-04-5 | C7H15N2O4P | 222.181 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. ionic liquid 1,3-dimethylimidazolium dimethylphosphate ([MMIM][DMP]) + water/ethanol/methanol mixtures exhibit properties which render them suitable as candidates for working pairs in industrial applications of absorption heat pumps or chillers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. ionic liquid 1,3-dimethylimidazolium dimethylphosphate ([MMIM][DMP]) + water/ethanol/methanol mixtures exhibit properties which render them suitable as candidates for working pairs in industrial applications of absorption heat pumps or chillers.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 654058-04-5
Molecular Formula C7H15N2O4P
Molecular Weight (g/mol) 222.181
MDL Number MFCD08275369
InChI Key GSGLHYXFTXGIAQ-UHFFFAOYSA-M
Synonym 1,3-dimethylimidazolium dimethyl phosphate, acmc-1b6nn, 1,3-dimethylimidazoliumdimethylphosphate, dsstox_cid_27946, dsstox_rid_82698, dsstox_gsid_47970, 1-methyl-3-methylimidazolium dimethylphosphate, 1,3-dimethyl-imidazolium dimethylphosphate, 1,3-dimethylimidazolium dimethyl phosphate hplc
PubChem CID 16069365
IUPAC Name 1,3-dimethylimidazol-1-ium;dimethyl phosphate
SMILES CN1C=C[N+](=C1)C.COP(=O)([O-])OC

Specifications

Density 1.277
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 222.18
Percent Purity 98%
Chemical Name or Material 1,3-Dimethylimidazolium dimethyl phosphate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.