Learn More
1,3-Dimethylimidazolium Methyl Sulfate 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Specifications
Specifications
| Quantity | 25 g |
| Synonym | 1,3-dimethylimidazolium methylsulfate, 1,3-dimethylimidazolium methyl sulfate, 1,3-dimethyl-1h-imidazol-3-ium methyl sulfate, 1,3-dimethylimidazolium methyl sulfate 1-, dmimmeso4, 1, 3-dimethylimidazolium methylsulfate, 1,3-dimethylimidazolium methyl sulfate, for catalysis hplc |
| Formula Weight | 208.23 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Liquid |
| Chemical Name or Material | 1,3-Dimethylimidazolium Methyl Sulfate |
Safety and Handling
| TSCA | No |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.