Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dimethylthiourea 97.0+%, TCI America™
SDP

Catalog No. D0804100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0804100G 100 g
D080425G 25 g
D0804500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0804100G Supplier TCI America Supplier No. D0804100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 534-13-4
Molecular Formula C3H8N2S
Molecular Weight (g/mol) 104.171
MDL Number MFCD00004923
InChI Key VLCDUOXHFNUCKK-UHFFFAOYSA-N
Synonym n,n'-dimethylthiourea, 1,3-dimethyl-2-thiourea, thiourea, n,n'-dimethyl, sym-dimethylthiourea, dimethylthiocarbamide, dimethylthiourea, 1,3-dimethylisothiourea, dimethyl-2-thiourea, urea, 1,3-dimethyl-2-thio, unii-8p30pmd17w
PubChem CID 2723631
IUPAC Name 1,3-dimethylthiourea
SMILES CNC(=S)NC

Specifications

Melting Point 65°C
Color White
Quantity 100 g
Formula Weight 104.17
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dimethylthiourea
EINECSNumber (2)-3266
RTECSNumber YT1520000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.