Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dimethyluracil 98.0+%, TCI America™
SDP

Catalog No. D08081G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D08081G 1 g
D08085G 5 g
D080825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D08081G Supplier TCI America Supplier No. D08081G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 874-14-6
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.14
MDL Number MFCD00038065
InChI Key JSDBKAHWADVXFU-UHFFFAOYSA-N
Synonym 1,3-dimethyluracil, n1,n3-dimethyluracil, n,n'-dimethyluracil, 1,3-dimethylpyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 1,3-dimethyl, 2,4-dihydroxy-1,3-dimethylpyrimidine, uracil, 1,3-dimethyl, 1,3-dimethyl-2,4 1h,3h-pyrimidinedione, 1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione, 1,3-dimethyl-1h-pyrimidine-2,4-dione
PubChem CID 70122
ChEBI CHEBI:74763
IUPAC Name 1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES CN1C=CC(=O)N(C)C1=O

Specifications

Melting Point 125°C
Color White
Quantity 1 g
Formula Weight 140.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dimethyluracil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.