Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dioxan-2-one 98.0+%, TCI America™
SDP

Catalog No. D382125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D382125G 25 g
D38215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D382125G Supplier TCI America Supplier No. D382125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2453-03-4
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
MDL Number MFCD00010440
InChI Key YFHICDDUDORKJB-UHFFFAOYSA-N
Synonym Trimethylene Carbonate
PubChem CID 123834
IUPAC Name 1,3-dioxan-2-one
SMILES C1COC(=O)OC1

Specifications

Melting Point 48°C
Color White
Boiling Point 135°C
Quantity 25 g
Formula Weight 102.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dioxan-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.