Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dioxane 98.0+%, TCI America™
SDP

Catalog No. D0859100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0859100ML 100 mL
D085925ML 25 mL
D0859500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0859100ML Supplier TCI America Supplier No. D0859100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 505-22-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00006566
InChI Key VDFVNEFVBPFDSB-UHFFFAOYSA-N
Synonym m-dioxane, 1,3-dioxacyclohexane, meta-dioxane, 1,3-propanediol formal, m-dioxin, dihydro, trimethylene glycol methylene ether, dihydro-m-dioxin, unii-b2c8m17i09, ccris 5911, 2,4-dioxane
PubChem CID 10450
ChEBI CHEBI:46924
IUPAC Name 1,3-dioxane
SMILES C1COCOC1

Specifications

Color Colorless
Boiling Point 105°C
Quantity 100 mL
UN Number 1165
Formula Weight 88.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dioxane
RTECSNumber JG8224000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.