Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dioxolane (stabilized with Triethylamine) 98.0+%, TCI America™
SDP

Catalog No. D0861500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0861500ML 500 mL
D086125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0861500ML Supplier TCI America Supplier No. D0861500ML

Chemical Identifiers

CAS 646-06-0
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.079
MDL Number MFCD00003207
InChI Key WNXJIVFYUVYPPR-UHFFFAOYSA-N
Synonym dioxolane, glycolformal, formal glycol, 1,3-dioxolan, 1,3-dioxacyclopentane, 1,3-dioxalane, ethylene glycol formal, glycol methylene ether, glycol formal, dioxolan czech
PubChem CID 12586
ChEBI CHEBI:87597
IUPAC Name 1,3-dioxolane
SMILES C1COCO1

Specifications

Color Yellow
Boiling Point 74°C
Quantity 500 mL
UN Number 1166
Formula Weight 74.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dioxolane (stabilized with Triethylamine)
EINECSNumber (5)-0500
RTECSNumber JH6760000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.