Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diphenyl-2-propanone 97.0+%, TCI America™
SDP

Catalog No. D0911100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0911100G 100 g
D091125G 25 g
D0911500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0911100G Supplier TCI America Supplier No. D0911100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 102-04-5
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.28
MDL Number MFCD00004795
InChI Key YFKBXYGUSOXJGS-UHFFFAOYSA-N
Synonym 1,3-diphenylacetone, dibenzyl ketone, 1,3-diphenyl-2-propanone, benzyl ketone, 2-propanone, 1,3-diphenyl, 1,3-diphenylpropanone, alpha,alpha'-diphenylacetone, 1,3-diphenyl-propan-2-one, unii-9y07g5udkq, fema no. 2397
PubChem CID 7593
IUPAC Name 1,3-diphenylpropan-2-one
SMILES O=C(CC1=CC=CC=C1)CC1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 100 g
Formula Weight 210.28
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,3-Diphenyl-2-propanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.