Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Hydroxybenzyl)piperidine 98.0+%, TCI America™
SDP

Catalog No. H118325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H118325G 25 g
H11835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H118325G Supplier TCI America Supplier No. H118325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 73279-04-6
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.274
MDL Number MFCD00800250
InChI Key ORGBERFQYFWYGX-UHFFFAOYSA-N
Synonym 3-1-piperidinylmethyl phenol, 3-piperidin-1-ylmethyl phenol, 3-1-piperidinomethyl phenol, 1-3-hydroxybenzyl piperidine, 3-piperidinomethyl phenol, 3-1-piperidylmethyl phenol, phenol, 3-1-piperidinylmethyl, 1-3-hydroxyphenylmethyl piperidine, pubchem6815, 3-piperidinomethylphenol
PubChem CID 826974
IUPAC Name 3-(piperidin-1-ylmethyl)phenol
SMILES C1CCN(CC1)CC2=CC(=CC=C2)O

Specifications

Melting Point 139°C
Color White-Yellow
Quantity 25 g
Formula Weight 191.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(3-Hydroxybenzyl)piperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.