Learn More
Sigma Organic Chemistry 1- 3- Methylthio phenyl | 1G | MFCD11841205
Supplier: Sigma Organic Chemistry CBR010141G

1- 3- Methylthio phenyl , 1G
About this Item:
MDL #: MFCD11841205
Molecular Weight: 189.71
UNSPSC Code: 12352200
Chemical Formula: C8H12ClNS
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.