Learn More
Medchemexpress LLC 1,3-propane-dithiol-d6 | 1219803-51-6 | 98.3% | 114.26 | C3H2D6S2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY151182S10MG
Encompass_Preferred
1,3-Propane-dithiol-d6 is a deuterium-labeled small molecule used as a tracer and internal standard for quantitative analytical techniques. It is supplied as a liquid for research use only, with manufacturer documentation providing storage, handling, and analytical report options. The compound is intended to support precise quantitation and method development in trace analysis workflows.
  • Deuterium-labeled internal standard for accurate quantitation.
  • Suitable as a tracer in analytical workflows.
  • Compatible with NMR, GC-MS, and LC-MS analyses.
  • Available in small-mass packages appropriate for trace-level work.
  • Storage stability under recommended cold conditions.
  • Datasheet and analytical reports available on request.
Catalog No. 50-002-85929
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.