Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Propane sultone, 97%

Catalog No. AC131420250
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC131420250 25 g
AC131421000 100 g
AC131420010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC131420250 Supplier Thermo Scientific Chemicals Supplier No. 131420250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 1120-71-4 | C3H6O3S | 122.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-71-4
Molecular Formula C3H6O3S
Molecular Weight (g/mol) 122.14
MDL Number MFCD00005355
InChI Key FSSPGSAQUIYDCN-UHFFFAOYSA-N
Synonym 1,3-propanesultone, propane sultone, 1,2-oxathiolane 2,2-dioxide, 1,3-propane sultone, 1,2-oxathiolane, 2,2-dioxide, propanesultone, rcra waste number u193, .gamma.-propane sultone, 3-hydroxy-1-propanesulfonic acid gamma-sultone, unii-l6ntk7vjx9
PubChem CID 14264
ChEBI CHEBI:82370
IUPAC Name oxathiolane 2,2-dioxide
SMILES O=S1(=O)CCCO1

Specifications

Melting Point 30.0°C to 33.0°C
Density 1.3920g/mL
Boiling Point 180.0°C (30.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25 g
Specific Gravity 1.392
Formula Weight 122.14
Percent Purity 97%
Chemical Name or Material 1,3-Propane sultone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
May cause cancer.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
EINECSNumber 214-317-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.