Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Propane sultone, MP Biomedicals™

Catalog No. p-4405209 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15639125 25 g
ICN15639150 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15639125 Supplier MP Biomedicals, Inc Supplier No. 0215639125
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1120-71-4
Molecular Formula C3H6O3S
Molecular Weight (g/mol) 122.14
MDL Number MFCD00005355
InChI Key FSSPGSAQUIYDCN-UHFFFAOYSA-N
Synonym 1,3-propanesultone, propane sultone, 1,2-oxathiolane 2,2-dioxide, 1,3-propane sultone, 1,2-oxathiolane, 2,2-dioxide, propanesultone, rcra waste number u193, .gamma.-propane sultone, 3-hydroxy-1-propanesulfonic acid gamma-sultone, unii-l6ntk7vjx9
PubChem CID 14264
ChEBI CHEBI:82370
IUPAC Name 1,2λ⁶-oxathiolane-2,2-dione
SMILES O=S1(=O)CCCO1

Specifications

Density 1.392g/mL at 25°C
Quantity 25 g
Formula Weight 122.14
Percent Purity 0.998
Physical Form Semi-solid
Chemical Name or Material 1, 3-Propane sultone, Cell Biology and Analysis
EINECSNumber 214-317-9
Recommended Storage Store at +4°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.